Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-17B (CHO) Protein, CF

Measured by its ability to induce IL-8 secretion by HepG2 human hepatocellular carcinoma cells. ED50 for this effect is 0.4 to 2 µg/mL.

$177.50 - $723.50

Specifications

Accession Number Q9UHF5
Buffer Lyophilized from a 0.2 µm filtered solution in Sodium Phosphate and NaCl.
Conjugate Unconjugated
Endotoxin Level <0.10 EU per 1 µg of the protein by the LAL method.
For Use With (Application) Bioactivity
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
81-29I-L010CF
View Documents
R&D Systems™
8129IL010/CF
10 µg
Each for $177.50
Add to cart
 
81-29I-L100CF
View Documents
R&D Systems™
8129IL100/CF
100 µg
Each for $723.50
Add to cart
 
Description

Description

Specifications

Specifications

Q9UHF5
Unconjugated
Bioactivity
18 kDa (monomer)
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Chinese Hamster Ovary cell line, CHO-derived human IL-17B protein Gln21-Phe180
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Lyophilized from a 0.2 µm filtered solution in Sodium Phosphate and NaCl.
<0.10 EU per 1 µg of the protein by the LAL method.
27190
Cytokine Zcyto7, cytokine-like protein ZCYTO7, IL17B, IL-17Binterleukin-17 beta, IL20, IL-20interleukin-17B, interleukin 17B, interleukin 20, Interleukin-20, MGC138900, MGC138901, neuronal interleukin-17 related factor, Neuronal interleukin-17-related factor, NIRF, ZCYTO7interleukin-20
Reconstitute at 100 µg/mL in PBS.
Human
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit