Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human IL-19 Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP2762022UG
Description
- An un-tagged recombinant protein corresponding to Human IL-19
- Amino Acid Sequence: MLRRCLISTD MHHIEESFQE IKRAIQAKDT FPNVTILSTL ETLQIIKPLD VCCVTKNLLA FYVDRVFKDH QEPNPKILRK ISSIANSFLY MQKTLRQCQE QRQCHCRQEA TNATRVIHDN YDQLEVHAAA IKSLGELDVF LAWINKNHEV MFSA
- Produced with no materials of animal or human origin using ANIMAL-FREE purification processes.
- No animal-derived ingredients, No FBS
- E. coli based production system
- Animal-Free facility, fermentor, columns, filters, labware, lyophilization, and storage
- Endotoxin levels at <1 EUs/ug
- Purity levels at 95% or greater
Specifications
Lyoph | |
29949 | |
>95%, by SDS-PAGE | |
E. coli | |
IL19 | |
Sterile water at 0.1 mg/mL |
SDS-PAGE | |
17.9 kDa | |
2 μg | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
IL-19 is a monomer, 17.9 kDa (154 aa) | |
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction