Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-2 (Cys145Ser) Protein, CF

Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells. ED50 for this effect is 0.05 to 0.25 ng/mL

$114.50 - $356.00

Specifications

Accession Number Q6NZ93
Buffer Supplied as a 0.2 µm filtered solution in Sodium Acetate.
Conjugate Unconjugated
Endotoxin Level <0.10 EU per 1 µg of the protein by the LAL method.
For Use With (Application) Bioactivity
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
10-81I-L010CF
View Documents
R&D Systems™
1081IL010/CF
10 µg
Each for $114.50
Add to cart
 
10-81I-L100CF
View Documents
R&D Systems™
1081IL100/CF
100 µg
Each for $356.00
Add to cart
 
Description

Description

Specifications

Specifications

Q6NZ93
Unconjugated
Bioactivity
15.5 kDa
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Human
Supplied as a 0.2 µm filtered solution in Sodium Acetate.
<0.10 EU per 1 µg of the protein by the LAL method.
3558
aldesleukin, IL2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF
E. coli-derived human IL-2 protein Ala21-Thr153 (Cys145Ser), with and without an N-terminal Met
6 months from date of receipt, 2 to 8 °C as supplied.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit