Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human IL-23 His Protein
Supplier: Novus Biologicals™ NBP280388250UG
Specifications
1 mg/mL | |
51561 | |
Baculovirus | |
< 1.0 EU per 1 microgram of protein (determined by LAL method) | |
PBS (pH 7.4), 10% glycerol |
SDS-PAGE | |
250 μg | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Human | |
> 95%, by SDS-PAGE |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction