Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IL-23 His Protein

$293.00 - $1049.00

Specifications

Buffer PBS (pH 7.4), 10% glycerol
Concentration 1 mg/mL
Endotoxin Concentration < 1.0 EU per 1 microgram of protein (determined by LAL method)
For Use With (Application) SDS-Page
Gene ID (Entrez) 51561
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB013460
View Documents
Novus Biologicals™
NBP28038850UG
50 μg
Each for $293.00
Add to cart
 
NB013459
View Documents
Novus Biologicals™
NBP280388250UG
250 μg
Each for $1,049.00
Add to cart
 
Description

Description

Specifications

Specifications

PBS (pH 7.4), 10% glycerol
< 1.0 EU per 1 microgram of protein (determined by LAL method)
51561
> 95%, by SDS-PAGE
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
1 mg/mL
SDS-Page
Human
Baculovirus
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit