Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human IL-4 (CHO-expressed) Protein
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. ED50 for this effect is 0.05 to 0.2 ng/mL.
$340.50 - $982.00
Specifications
Accession Number | P05112 |
---|---|
For Use With (Application) | Bioactivity |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
Gene ID (Entrez) | 3565 |
Molecular Weight (g/mol) | M.W. (Observed): 17-25 kDa, reducing conditions; M.W. (theoretical): 15.0 kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
65-07I-L025CF
|
R&D Systems™
6507IL025/CF |
25 μg |
Each for $340.50
|
|
|||||
65-07I-L100CF
|
R&D Systems™
6507IL100/CF |
100 μg |
Each for $982.00
|
|
|||||
Description
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is 0.05-0.2 ng/mL.Specifications
P05112 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
M.W. (Observed): 17-25 kDa, reducing conditions; M.W. (theoretical): 15.0 kDa | |
Chinese Hamster Ovary cell line, CHO-derived human IL-4 protein His25-Ser153 | |
<0.01 EU / 1 μg of the protein by the LAL method. | |
IL4 | |
Human | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Bioactivity | |
3565 | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
Unconjugated | |
Reconstitute at 100 μg/mL in PBS. | |
<0.01 EU per 1 μg of the protein by the LAL method. | |
IL-4 |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title