Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human IL-5 Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$133.00 - $358.00

Specifications

Source Baculovirus
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol IL5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25951310 View Documents Novus Biologicals™
NBP25951310UG
10μg Each for $133.00
Add to cart
 
NBP25951350 View Documents Novus Biologicals™
NBP25951350UG
50μg Each for $358.00
Add to cart
 
Description

Description

A bioactive recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 20-134 of Human IL-5 The Recombinant Human IL-5 Protein is derived from E. coli. The Recombinant Human IL-5 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

Baculovirus
RUO
IL5
3567
Recombinant Human IL-5 Protein
Liquid. PBS (pH 7.4) containing 10% glycerol
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
B-cell differentiation factor I, EDF, Eosinophil differentiation factor, IL-5T-cell replacing factor, interleukin 5 (colony-stimulating factor, eosinophil), interleukin-5, TRFB cell differentiation factor I
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
14.2 kDa
< 1.0 EU per 1 microgram of protein (determined by LAL method)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit