Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human IL-6 (HEK293-expressed) Protein, CF

Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. ED50 for this effect is 0.1 to 0.6 ng/mL

$177.50 - $702.50

Specifications

Accession Number P05231
Buffer Lyophilized from a 0.2 µm filtered solution in PBS.
Conjugate Unconjugated
Endotoxin Level <0.10 EU per 1 µg of the protein by the LAL method.
For Use With (Application) Bioactivity
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
72-70I-L010CF
View Documents
R&D Systems™
7270IL010/CF
10 µg
Each for $177.50
Add to cart
 
72-70I-L100CF
View Documents
R&D Systems™
7270IL100/CF
100 µg
Each for $702.50
Add to cart
 
Description

Description

Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.1-0.6 ng/mL.
Specifications

Specifications

P05231
Unconjugated
Bioactivity
M.W. (Observed): 22 kDa, reducing conditions; M.W. (theoretical): 20.8 kDa
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Human embryonic kidney cell, HEK293-derived human IL-6 protein Met1-Met212
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Lyophilized from a 0.2 µm filtered solution in PBS.
<0.10 EU per 1 µg of the protein by the LAL method.
3569
B cell stimulatory factor-2, B-cell differentiation factor, BSF2, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Reconstitute at 100 µg/mL in PBS.
Human
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit