Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Integrin beta 1/CD29 Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$281.00 - $1004.00

Specifications

Source Baculovirus
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol ITGB1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25957050 View Documents Novus Biologicals™
NBP25957050UG
50μg Each for $281.00
Add to cart
 
NBP259570Y View Documents Novus Biologicals™
NBP259570250UG
250μg Each for $1,004.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-728 of Human Integrin beta 1/CD29 The Recombinant Human Integrin beta 1/CD29 Protein is derived from E. coli. The Recombinant Human Integrin beta 1/CD29 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

Baculovirus
RUO
ITGB1
3688
Recombinant Human Integrin beta 1/CD29 Protein
Liquid. PBS (pH 7.4) containing 10% glycerol
Unlabeled
1mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, b
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
79.4 kDa
<1.0 EU per 1μg of protein (determined by LAL method)

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit