Learn More
Novus Biologicals™ Recombinant Human Intelectin-1/Omentin Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP2763161MG
 View more versions of this product
Catalog No. NBP2763161M
Add to cart
Description
Description
An un-tagged recombinant protein corresponding to Human Intelectin-1/Omentin
The Recombinant Human Intelectin-1/Omentin Animal-Free Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Lyoph
55600
>90%, by SDS-PAGE
E. coli
ITLN1
Sterile water at 0.1 mg/mL
SDS-Page
35 kDa
1 mg
Store at -20 to -80°C. Avoid freeze-thaw cycles.
Intelectin-1/Omentin is a monomer, 35 kDa (313 aa)
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate and 5:1 mannitol to protein, pH 7.5