Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Recombinant Human Intelectin-1/Omentin Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
$83.90 - $5128.40
Specifications
Buffer | Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate and 5:1 mannitol to protein, pH 7.5 |
---|---|
Concentration | Lyoph |
For Use With (Application) | SDS-Page |
Gene ID (Entrez) | 55600 |
Gene Symbol | ITLN1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2763052U
|
Novus Biologicals
NBP2763052UG |
2 μg |
Each for $83.90
|
|
NB27630510U
|
Novus Biologicals
NBP27630510UG |
10 μg |
Each for $190.10
|
|
NBP2763051M
|
Novus Biologicals
NBP2763051MG |
1 mg |
Each for $5,128.40
|
|
Description
An un-tagged recombinant protein corresponding to Human Intelectin-1/OmentinThe Recombinant Human Intelectin-1/Omentin Protein has been validated for the following applications: SDS-Page.
Specifications
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate and 5:1 mannitol to protein, pH 7.5 | |
SDS-Page | |
ITLN1 | |
>90%, by SDS-PAGE | |
E. coli | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. |
Lyoph | |
55600 | |
35 kDa | |
Sterile water at 0.1 mg/mL | |
Intelectin-1/Omentin is a monomer, 35 kDa (313 aa) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title