Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Kallikrein 15 Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$398.00 - $1472.00
Specifications
Concentration | 0.25mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. PBS (pH 7.4) containing 10% glycerol |
Gene ID (Entrez) | 55554 |
Molecular Weight (g/mol) | 27.4 kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25959510
|
Novus Biologicals™
NBP25959510UG |
10μg |
Each for $398.00
|
|
|||||
NBP25959550
|
Novus Biologicals™
NBP25959550UG |
50μg |
Each for $1,472.00
|
|
|||||
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 17-256 of Human Kallikrein 15 The Recombinant Human Kallikrein 15 Protein is derived from E. coli. The Recombinant Human Kallikrein 15 Protein has been validated for the following applications: SDS-Page.Specifications
0.25mg/mL | |
Liquid. PBS (pH 7.4) containing 10% glycerol | |
27.4 kDa | |
>90%, by SDS-PAGE | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
<1.0 EU per 1μg of protein (determined by LAL method) | |
KLK15 | |
Unconjugated | |
Recombinant |
SDS-PAGE | |
55554 | |
Recombinant Human Kallikrein 15 Protein | |
Baculovirus | |
RUO | |
ACO, ACO protease, EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, HSRNASPH, kallikrein 15, kallikrein-15, kallikrein-like serine protease, kallikrein-related peptidase 15, prostinogen | |
Recombinant Protein | |
Human |
For Research Use Only.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title