Learn More
R&D Systems™ Recombinant Human LAIR1 Fc Chimera Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9964LR050
Catalog No. 99-64LR-050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human LAIR1 Fc Chimera Protein is derived from HEK293. The Recombinant Human LAIR1 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
NP_002278
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
42 kDa
50 μg
12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution.
<0.10 EU per 1 μg of the protein by the LAL method.
LAIR1
Reconstitute at 500 μg/mL in PBS.
Bioactivity
3903
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Human embryonic kidney cell, HEK293-derived human LAIR1 protein
RUO
Unconjugated
Human