Learn More
R&D Systems™ Recombinant Human LRFN5 Fc Chimera Protein CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9385SA050
Catalog No. 93-85S-A050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human LRFN5 Fc Chimera Protein is derived from HEK293. The Recombinant Human LRFN5 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Q96NI6
Bioactivity
145581
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
HEK293
<0.10EU per 1μg of the protein by the LAL method.
Recombinant Protein
Human
Bioactivity
Lyophilized from a 0.2μm filtered solution in PBS.
83kDa
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
C14orf146, chromosome 14 open reading frame 146, DKFZp686G0210, fibronectin type III, immunoglobulin and leucine rich repeat domains 8, FIGLER8, leucine rich repeat and fibronectin type III domain containing 5, leucine-rich repeat and fibronectin type-III domain-containing protein 5, SALM5, SALM5FLJ30803
Unconjugated
Reconstitute at 500μg/mL in PBS.