Learn More
R&D Systems™ Recombinant Human MAN2B1 Protein CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9826GH050
Catalog No. 98-26G-H050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human MAN2B1 Protein is derived from HEK293. The Recombinant Human MAN2B1 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
O00754-1
Supplied as a 0.2μm filtered solution in Tris and NaCl.
110 kDa
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
HEK293
<0.10 EU per 1μg of the protein by the LAL method.
Human
Enzyme Activity
4125
alpha B, lysosomal, EC 3.2.1, EC 3.2.1.24, LAMAN, Lysosomal acid alpha-mannosidase, MANB, Mannosidase alpha class 2B member 1, Mannosidase Alpha-B, mannosidase, alpha, class 2B, member 1
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Unconjugated