Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Methionine Aminopeptidase 1/METAP1 Protein

Click to view available options
Quantity:
100μg
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-386 of Human Methionine Aminopeptidase 1/METAP1 The Recombinant Human Methionine Aminopeptidase 1/METAP1 Protein is derived from E. coli. The Recombinant Human Methionine Aminopeptidase 1/METAP1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
Concentration | 1mg/mL |
Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 3mM DTT, 40% glycerol, 200mM NaCl, 0.1mM PMSF |
Gene ID (Entrez) | 23173 |
Quantity | 100μg |
Storage Requirements | Store at 4° short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Regulatory Status | RUO - research use only |
Species | Human |
Recombinant | Recombinant |
Purity or Quality Grade | >90%, by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction