Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human MICB His-tag Protein
Click to view available options
Quantity:
50 μg
Description
When Recombinant Human NKG2D/CD314 Fc Chimera (Catalog # 1299-NK ) is immobilized at 0.125 μg/mL, 100 μL/well, the concentration of Recombinant Human MICB His-tag (Catalog # 10431-MB) that produces 50% of the optimal binding response is approximately 80-400 ng/mL.
Specifications
Specifications
Accession Number | NP_005922.2 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
Gene ID (Entrez) | 4277 |
Molecular Weight (g/mol) | Observed: 35-55 kDa, under reducing conditions; Theroretical: 33 kDa |
Quantity | 50 μg |
Source | Chinese Hamster Ovary cell line, CHO-derived human MICB protein Ala23-Thr308, with a C-terminal 6-His tag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
Conjugate | Unconjugated |
Reconstitution | Reconstitute at 500 μg/mL in PBS. |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction