Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat BMP-2 (E. coli-expressed), CF

Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. ED50 for this effect is 0.08 to 0.48 µg/mL.

$356.00 - $839.00

Specifications

Accession Number P12643
Buffer Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA.
Conjugate Unconjugated
Endotoxin Level <0.01 EU per 1 µg of the protein by the LAL method.
For Use With (Application) Bioactivity
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
35-5BE-C025CF
View Documents
R&D Systems™
355BEC025/CF
25 µg
Each for $356.00
Add to cart
 
35-5BE-C100CF
View Documents
R&D Systems™
355BEC100/CF
100 µg
Each for $839.00
Add to cart
 
Description

Description

Specifications

Specifications

P12643
Unconjugated
Bioactivity
12.8 kDa (monomer)
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain
E. coli-derived BMP-2 protein Ala284-Arg396 This form is not detected using human BMP-2 Quantikine ELISA (Catalog # DBP200 ).
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA.
<0.01 EU per 1 µg of the protein by the LAL method.
650
BDA2, BMP2, BMP-2, BMP-2A, BMP2ABone morphogenetic protein 2A, bone morphogenetic protein 2, SSFSC
Reconstitute at 100 µg/mL in sterile 4 mM HCl.
Human, Mouse, Rat
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit