Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat GDF-8/Myostatin Protein

Measured by its ability to induce hemoglobin expression in K562 human chronic myelogenous leukemia cells. The ED50 for this effect is 2 to 10 ng/mL.

$1259.00 - $2050.00

Specifications

Accession Number O08689
Buffer Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 µm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
Conjugate Unconjugated
Endotoxin Level <0.10 EU per 1 µg of the protein by the LAL method.
For Use With (Application) Bioactivity
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
78-8G8-100
View Documents
R&D Systems™
788G8100
100 µg
Each for $1,259.00
Add to cart
 
78-8G8-250
View Documents
R&D Systems™
788G8250
250 µg
Each for $2,050.00
Add to cart
 
Description

Description

Specifications

Specifications

O08689
Unconjugated
Bioactivity
12.4 kDa
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Mouse myeloma cell line, NS0-derived GDF-8/Myostatin protein Asp268-Ser376
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. *1 mg pack size (01M) is supplied as a 0.2 µm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
<0.10 EU per 1 µg of the protein by the LAL method.
2660
GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Reconstitute at 100 µg/mL in sterile 4 mM HCl containing at least 0.1% human or bovine serum albumin.
Human, Mouse, Rat
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit