Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human MuSK His-tag Protein, CF
Supplier: R&D Systems™ 10189MK050
This item is not returnable.
View return policy
Description
Measured by its binding ability in a functional ELISA. In the presence of Recombinant Human Agrin (Catalog # 6624-AG ), immobilized Recombinant Human MuSK His-tag protein (2 μg/mL at 100 μL/well) binds Recombinant Human LRP-4 (Catalog # 5948-LR ) with an ED50 of 0.75-4.5 μg/mL.Specifications
O15146.1 | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. | |
M.W.-observed: 62-75 kDa, under reducing conditions, M.W.-theroretical: 53 kDa | |
50 µg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
EC 2.7.10, EC 2.7.10.1, MGC126323, MGC126324, muscle, skeletal, receptor tyrosine kinase, MuSK, skeletal receptor tyrosine-protein kinase | |
Reconstitute at 500 µg/mL in PBS. |
Bioactivity | |
4593 | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. | |
Chinese Hamster Ovary cell line, CHO-derived human MuSK protein Leu24-Thr495, with a C-terminal 6-His tag | |
<0.10 EU / 1 µg of the protein by the LAL method. | |
Unconjugated | |
MuSK |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title