Learn More
R&D Systems™ Recombinant Human Ninjurin-1 Protein CF
R&D Systems™ Recombinant Human Ninjurin-1 Protein CF is a member of the Ninjurin family of transmembrane (TM) proteins.
Manufacturer:  R&D Systems™ 9717NJ050
Catalog No. 9717NJ050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human Ninjurin-1 Protein is derived from E. coli.
Specifications
Specifications
Q92982
Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
9 kDa
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli
<1.0 EU per 1μg of the protein by the LAL method.
Human
Bioactivity not tested
4814
Nerve injury-induced protein 1, nerve injury-induced protein-1, NIN1, NINJ1, NINJURIN, ninjurin 1, ninjurin-1, Ninjurin1
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Reconstitute at 250μg/mL in PBS.