Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human Notum Protein

Catalog No. 9118NM050
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Notum Protein is derived from CHO. The Recombinant Human Notum Protein has been validated for the following applications: Bioactivity.

Specifications

Accession Number Q6P988
For Use With (Application) Bioactivity
Gene ID (Entrez) 147111
Molecular Weight (g/mol) Observed M.W.: 38-45kDa, reducing conditions, Theoretical M.W.: 43kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Notum protein Ser81-Thr451 (Cys330Ser) with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias EC 3.-, notum pectinacetylesterase homolog (Drosophila), protein notum homolog
Common Name NOTUM
Biological Activity Measured by its ability to inhibit Wnt induced TCF reporter activity in HEK293 human embryonic kidney cells. The ED50 for this effect is 0.1-0.6μg/mL in the presence of 0.5μg/mL of Recombinant Human Wnt-3a.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Ser81-Thr451 (Cys330Ser) with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.