Learn More
R&D Systems™ Recombinant Human PARP Enzyme (HSA)
R&D Systems™ Recombinant Human PARP Enzyme (HSA) serves as a measure of enzyme purity, with greater than 95% purity for PARP-HSA.
Manufacturer:  R&D Systems™ 466802K01
Catalog No. 46-680-2K01
Add to cart
This item is not returnable. View return policy
Description
Description
  • PARP1 is activated by 10X Activated DNA
  • PARP1 uses NAD+ as a substrate to catalyze poly(ADP-ribosyl)ation of target proteins involved in chromatin structure and DNA metabolism. PARP1 is stimulated by DNA-strand breaks and capable of auto-ribosylation.
  • For identification of inhibitors of PARP1 activity
  • For quantitation of DNA Damage
  • For investigation of PARP1 inactivation during apoptosis
  • ForWestern Blot Analysis
Specifications
Specifications
10 units/μL
20000 Units
Store the unopened product at -70°C
Purified from E. coli containing a recombinant plasmid harboring human PARP1 gene.
PARP Assay Kits