Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Recombinant Human PDGF-AA, Animal-Free Protein

Measured in a cell proliferation assay using NR6R-3T3 mouse fibroblast cells. ED50 for this effect is 50 to 200 ng/mL in a fluorometric assay using the redox sensitive dye, Resazurin.

$303.50 - $944.00

Specifications

Accession Number P04085
Buffer Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA.
Conjugate Unconjugated
Endotoxin Level <0.01 EU per 1 µg of the protein by the LAL method.
For Use With (Application) Bioactivity
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AFL221010
View Documents
R&D Systems™
AFL221010
10 µg
Each for $303.50
Add to cart
 
AFL221050
View Documents
R&D Systems™
AFL221050
50 µg
Each for $944.00
Add to cart
 
Description

Description

Specifications

Specifications

P04085
Unconjugated
Bioactivity
M.W. theroretical: 14 kDa (monomer)
Reconstitute at 0.2 mg/mL in sterile 4 mM HCl.
Human
Lyophilized from a 0.2 µm filtered solution in Acetonitrile and TFA.
<0.01 EU per 1 µg of the protein by the LAL method.
5154
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
E. coli-derived human PDGF-AA protein Ser87-Thr211, with and without an N-terminal Met Produced using non-animal reagents in an animal-free laboratory.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, ≤ -20°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit