Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human PHPT1 Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$129.70 - $351.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol PHPT1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25958120 View Documents Novus Biologicals™
NBP25958120UG
20μg Each for $129.70
Add to cart
 
NBP259581X View Documents Novus Biologicals™
NBP259581100UG
100μg Each for $351.00
Add to cart
 
Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-125 of Human PHPT1 The Recombinant Human PHPT1 Protein is derived from E. coli. The Recombinant Human PHPT1 Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

E.coli
RUO
PHPT1
29085
Recombinant Human PHPT1 Protein
Liquid. 20 mM Tris-HCl buffer (pH8.0) containing 0.2M NaCl, 2mM DTT, 10% glycerol
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein jan
Recombinant Protein
>90%, by SDS-PAGE
Functional, SDS-Page
15.9 kDa
Human

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit