Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human Prolactin Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP27623250UG
Description
An un-tagged bioactive recombinant protein corresponding to Human ProlactinThe Recombinant Human Prolactin Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Lyoph | |
5617 | |
>95%, by SDS-PAGE | |
E. coli | |
PRL | |
Sterile water at 0.1 mg/mL |
Bioactivity, SDS-PAGE | |
23 kDa | |
50 μg | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
Prolactin is a monomer, 23 kDa (200 aa) | |
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 10 mM sodium phosphate, pH 7.5 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction