Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human Proprotein Convertase 9/PCSK9 Protein
Click to view available options
Quantity:
20 μg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human LDLR (Catalog # 2148-LD/CF ) is immobilized at 1μg/mL (100μL/well), Biotinylated Recombinant Human Proprotein Convertase 9/PCSK9 His-tag Avi-tag (Catalog # AVI3888) binds with an ED50 of 0.150-1.20μg/mL.
Specifications
Specifications
Accession Number | ABV59216.1 |
For Use With (Application) | Bioactivity |
Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl and Glycerol. |
Gene ID (Entrez) | 255738 |
Molecular Weight (g/mol) | M.W. (Observed): 18-21 & 62-69, under reducing conditions, M.W. (Theoretical): 14 kDa & 60 kDa |
Quantity | 20 μg |
Source | Chinese Hamster Ovary cell line, CHO-derived human Proprotein Convertase 9/PCSK9 protein Gln31-Gln152 (pro domain), Ser153-Gln692 (mature form) with a C-terminal 6-His and Avitag |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. |
Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
Gene Alias | EC 3.4.21, EC 3.4.21.111, FH3, FH3neural apoptosis regulated convertase 1, FHCL3, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC1, NARC-1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, PCSK9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction