Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Protein Phosphatase 1C gamma Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$129.00 - $280.50

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol PPP1CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2595825 View Documents Novus Biologicals™
NBP2595825UG
5μg Each for $129.00
Add to cart
 
NBP25958220 View Documents Novus Biologicals™
NBP25958220UG
20μg Each for $280.50
Add to cart
 
Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-323 of Human Protein Phosphatase 1C gamma The Recombinant Human Protein Phosphatase 1C gamma Protein is derived from E. coli. The Recombinant Human Protein Phosphatase 1C gamma Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

E.coli
RUO
PPP1CC
5501
Recombinant Human Protein Phosphatase 1C gamma Protein
Liquid. 20mM Tris-HCl buffer (pH8.0) containing 50% glycerol, 0.2M NaCl, 2mM DTT
Unlabeled
0.25mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
EC 3.1.3.16, PP-1G, PP1gamma, PPP1G, protein phosphatase 1, catalytic subunit, gamma isoform, protein phosphatase 1, catalytic subunit, gamma isozyme, Protein phosphatase 1C catalytic subunit, serine/threonine phosphatase 1 gamma, serine/threonine-protein
Recombinant Protein
>90%, by SDS-PAGE
Functional, SDS-Page
39.1 kDa
Human

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit