Learn More
R&D Systems™ Recombinant Human PTK7/CCK4 Fc Chimera Protein, CF
Recombinant human protein
Manufacturer:  R&D Systems™ 9799TK050
Catalog No. 9799TK050
Add to cart
This item is not returnable. View return policy
Description
Description
Measured by its binding ability in a functional ELISA. In a 100 µL reaction mixture containing biotinylated Recombinant Mouse Wnt-3a at 50 ng/mL and Recombinant Human PTK7/CCK4 Chimera dilutions, the concentration of Recombinant Human PTK7/CCK4 Fc Chimera that produces 50% of the maximal binding response is 4-20 µg/mL.
Source:  Human embryonic kidney cell, HEK293-derived human PTK7/CCK4 protein
Specifications
Specifications
Q13308-1
5754
50 μg
RUO
Unconjugated
Reconstitute at 500 μg/ml in PBS.
Bioactivity
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
HEK293
<0.10 EU per 1μg of the protein by the LAL method.
PTK7/CCK4