Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals Recombinant Human RELM beta Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
$83.90 - $3163.70
Specifications
Buffer | Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA) |
---|---|
Concentration | Lyoph |
For Use With (Application) | SDS-Page |
Gene ID (Entrez) | 84666 |
Gene Symbol | RETNLB |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2763005U
|
Novus Biologicals
NBP2763005UG |
5 μg |
Each for $83.90
|
|
NB27630025U
|
Novus Biologicals
NBP27630025UG |
25 μg |
Each for $190.10
|
|
NBP2763001M
|
Novus Biologicals
NBP2763001MG |
1 mg |
Each for $3,163.70
|
|
Description
An un-tagged recombinant protein corresponding to Human RELM betaThe Recombinant Human RELM beta Animal-Free Protein has been validated for the following applications: SDS-Page.
Specifications
Lyophilized from a sterile (0.2 micron) filtered aqueous solution containing 0.1% Trifluoroacetic Acid (TFA) | |
SDS-Page | |
RETNLB | |
>90%, by SDS-PAGE | |
E. coli | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. |
Lyoph | |
84666 | |
19 kDa | |
Sterile 0.1% Trifluoroacetic Acid(TFA) at 0.1 mg/mL | |
RELM beta is a non-covalent homodimer, 9.5/19.0 kDa (89/178 aa) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title