Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Semaphorin 7A Isoform 1 Protein
SDP

Highly purified. Generating reliable and reproducible results.

$398.00 - $1472.00

Specifications

Concentration 0.25 mg/ml
For Use With (Application) SDS-PAGE
Formulation Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
Gene ID (Entrez) 8482
Molecular Weight (g/mol) 95.7 kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP26197710
SDP
View Documents
Novus Biologicals™
NBP26197710UG
10 μg
Each for $398.00
Add to Cart
 
NBP26197750
SDP
View Documents
Novus Biologicals™
NBP26197750UG
50 μg
Each for $1,472.00
Add to Cart
 
Description

Description

A recombinant protein with a N-Terminal hIgG-His-tag and corresponding to the amino acids 45-648 of Human Semaphorin 7A The Recombinant Human Semaphorin 7A Isoform 1 Protein is derived from Baculovirus. The Recombinant Human Semaphorin 7A Isoform 1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.25 mg/ml
Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
95.7 kDa
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
< 1.0 EU per 1μg of protein (determined by LAL method)
Semaphorin 7A
>90%, by SDS-PAGE
SDS-PAGE
8482
Baculovirus
RUO
SEMA7A
Human
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit