Learn More
Novus Biologicals™ Recombinant Human Serpin A5/Protein C Inhibitor Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity
Manufacturer:  Novus Biologicals™ NBP2619915UG
 View more versions of this product
Catalog No. NBP2619915
Add to cart
Description
Description
A Biologically active recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 20-406 of Human Serpin A8/Angiotensinogen The Recombinant Human Serpin A5/Protein C Inhibitor Protein is derived from E. coli. The Recombinant Human Serpin A5/Protein C Inhibitor Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
0.5 mg/ml
20 mM MES buffer (pH6.0) containing 150mM NaCl, 10% glycerol
45.9 kDa
5 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SERPINA5
Human
SDS-Page, Bioactivity
5104
>90%, by SDS-PAGE
E. coli
RUO
Serpin A5/Protein C Inhibitor