Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human SP-D Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

Manufacturer:  Novus Biologicals™ NBP26199250UG

 View more versions of this product

Catalog No. NBP26199250


Add to cart

Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 22-375 of Human SP-D The Recombinant Human SP-D Protein is derived from Baculovirus. The Recombinant Human SP-D Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

0.25 mg/ml
Phosphate Buffered Saline (pH 7.4) containing 10% glycerol.
36.5 kDa
50 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
< 1.0 EU per 1μg of protein (determined by LAL method)
SP-D
SDS-Page
6441
>90%, by SDS-PAGE
Baculovirus
RUO
SFTPD
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit