Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems Recombinant Human SPR His-tag Protein, CF
Measured by its ability to catalyze the reduction of phenanthrenequinone. The specific activity is >700 pmol/min/µg, as measured under the described conditions.
Manufacturer: R&D Systems 10209SP020
This item is not returnable.
View return policy
Specifications
P35270 | |
Human | |
6697 | |
EC 1.1.1.153, Gm10328, SDR38C1, sepiapterin reductase, sepiapterin reductase (7,8-dihydrobiopterin:NADP+ oxidoreductase), short chain dehydrogenase/reductase family 38C, member 1 | |
20 µg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70 °C as supplied. 3 months, -20 to -70 °C under sterile conditions after opening. | |
Supplied as a 0.2 µm filtered solution in Tris, NaCl and Glycerol. |
<0.10 EU per 1 µg of the protein by the LAL method. | |
Enzyme Activity | |
29 kDa | |
>90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. | |
E. coli-derived human SPR protein Met1-Lys261 with a C-terminal 6-His tag | |
Unconjugated |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title