Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human Sulfatase Modifying Factor 1/SUMF1 Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$366.50 - $1784.00

Specifications

Concentration 0.25 mg/mL
Endotoxin Level < 1.0 EU per 1 μg of protein (determined by LAL method)
Purity >85%, by SDS-PAGE
Source Baculovirus
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB002928
View Documents
Novus Biologicals™
NBP2774880.05MG
0.05 mg
Each for $366.50
Add to cart
 
NB002929
View Documents
Novus Biologicals™
NBP2774880.1MG
0.1 mg
Each for $576.50
Add to cart
 
NB002930
View Documents
Novus Biologicals™
NBP2774880.5MG
0.5 mg
Each for $1,784.00
Add to cart
 
Description

Description

A recombinant protein with a His-Tag and corresponding to the amino acids 34-374aa of Human Sulfatase Modifying Factor 1/SUMF1.

Source: Baculovirus

Amino Acid Sequence:

SQEAGTGAGA GSLAGSCGCG TPQRPGAHGS SAAAHRYSRE ANAPGPVPGE RQLAHSKMVP IPAGVFTMGT DDPQIKQDGE APARRVTIDA FYMDAYEVSN TEFEKFVNST GYLTEAEKFG DSFVFEGMLS EQVKTNIQQA VAAAPWWLPV KGANWRHPEG PDSTILHRPD HPVLHVSWND AVAYCTWAGK RLPTEAEWEY SCRGGLHNRL FPWGNKLQPK GQHYANIWQG EFPVTNTGED GFQGTAPVDA FPPNGYGLYN IVGNAWEWTS DWWTVHHSVE ETLNPKGPPS GKDRVKKGGS YMCHRSYCYR YRCAARSQNT PDSSASNLGF RCAADRLPTM DHHHHHH>

Specifications

Specifications

0.25 mg/mL
< 1.0 EU per 1 μg of protein (determined by LAL method)
>85%, by SDS-PAGE
Baculovirus
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit