Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human TDP-43/TARDBP Protein

Highly purified. Generating reliable and reproducible results.

$284.00 - $783.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol TARDBP
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25955110 View Documents Novus Biologicals™
NBP25955110UG
10μg Each for $284.00
Add to cart
 
NBP25955150 View Documents Novus Biologicals™
NBP25955150UG
50μg Each for $783.00
Add to cart
 
Description

Description

An un-tagged recombinant protein corresponding to the amino acids 1-414 of Human TDP-43/TARDBP The Recombinant Human TDP-43/TARDBP Protein is derived from E. coli. The Recombinant Human TDP-43/TARDBP Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

E.coli
RUO
TARDBP
23435
Recombinant Human TDP-43/TARDBP Protein
Liquid. 20mM Tris-HCl(pH8.0) containing 0.1M NaCl, 1mM DTT, 1mM EDTA, 10% glycerol
Unlabeled
0.25mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
ALS10TAR DNA-binding protein-43, TAR DNA binding protein, TDP43, TDP-43, TDP-43TAR DNA-binding protein 43
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
44.7 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit