Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human TGF-beta 3 Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP27623610UG
Description
An un-tagged bioactive recombinant protein corresponding to Human TGF-beta 3The Recombinant Human TGF-beta 3 Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
0.25 mg/mL | |
7043 | |
>95%, by SDS-PAGE | |
E. coli | |
TGFB3 | |
10 mM acetic acid and 20% Ethanol |
Bioactivity, SDS-PAGE | |
25.7 kDa | |
10 μg | |
Store at 4°C | |
TGF-beta 3 is a dimer, 12.9/25.7 kDa (113/226 aa) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction