Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Human TRAILR1/TNFRSF10A Fc Chimera Protein
Click to view available options
Quantity:
100 μg
Description
The Recombinant Human TRAIL R1/TNFRSF10A Fc Chimera Protein is derived from Sf 21 (baculovirus). The Recombinant Human TRAIL R1/TNFRSF10A Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
Gene ID (Entrez) | 8797 |
Molecular Weight (g/mol) | 43 kDa (monomer) |
Name | TRAILR1/TNFRSF10A |
Quantity | 100 μg |
Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human TRAIL R1/TNFRSF10A protein, (N-terminus) Human TRAIL R1 (Ala109-Asn239) & (Thr110-Asn239) Accession &Num; AAC51226 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus) |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Structural Form | Disulfide-linked homodimer |
Gene Alias | APO2, CD261, CD261 antigen, cytotoxic TRAIL receptor, Death receptor 4, DR4, DR4 TRAIL receptor 1, TNF-related apoptosis inducing ligand receptor 1, TNF-related apoptosis-inducing ligand receptor 1, TNFRSF10A, TRAIL R1, TRAIL-R1, TRAILR-1, TRAILR1MGC9365, tumor necrosis factor receptor superfamily member 10A, tumor necrosis factor receptor superfamily, member 10a |
Conjugate | Unconjugated |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction