Learn More
R&D Systems™ Recombinant Human TREM2 His-tag Protein, CF Available on GSA/VA Contract for Federal Government customers only.
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9256T2050
Catalog No. 92-56T-2050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human TREM-2 His-tag Protein is derived from NS0. The Recombinant Human TREM-2 His-tag Protein has been validated for the following applications: Bioactivity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
Bioactivity
TREM2
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
PLOSL2, TREM-2, Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2