Learn More
R&D Systems™ Recombinant Human TrkA His Tagged Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9966TK050
Catalog No. 99-66TK-050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human TrkA His Tagged Protein is derived from HEK293. The Recombinant Human TrkA His Tagged Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
P04629-1
Lyophilized from a 0.2 μm filtered solution in PBS.
43 kDa
50 μg
12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution.
<0.10 EU per 1 μg of the protein by the LAL method.
TrkA
Reconstitute at 1 mg/mL in PBS.
Bioactivity
4914
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Human embryonic kidney cell, HEK293-derived human TrkA protein Ala34-Gly423, with a C-terminal 6-His tag
RUO
Unconjugated
Human