Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Recombinant Human VEGF Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
$398.00 - $1110.00
Specifications
For Use With (Application) | SDS-PAGE |
---|---|
Formulation | Phosphate buffered saline (pH 7.4), 30% glycerol, 1mM DTT, 0.1mM PMSF |
Gene ID (Entrez) | 7422 |
Molecular Weight (g/mol) | 19.9kDa |
Name | Human VEGF Protein |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB25304020
|
Novus Biologicals™
NBP25304020UG |
20μg |
Each for $398.00
|
|
|||||
NB253040100
|
Novus Biologicals™
NBP253040100UG |
100μg |
Each for $1,110.00
|
|
|||||
Description
A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 1-191 of Human VEGF The Recombinant Human VEGF Protein is derived from E. coli. The Recombinant Human VEGF Protein has been validated for the following applications: SDS-Page.Specifications
SDS-PAGE | |
7422 | |
Human VEGF Protein | |
Baculovirus | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor | |
Recombinant Protein | |
Human |
Phosphate buffered saline (pH 7.4), 30% glycerol, 1mM DTT, 0.1mM PMSF | |
19.9kDa | |
>85%, by SDS-PAGE | |
APMAEGGGQN HHEVVKFMDV YQRSYCHPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCGGC CNDEGLECVP TEESNITMQI MRIKPHQGQH IGEMSFLQHN KCECRPKKDR ARQENPCGPC SERRKHLFVQ DPQTCKCSCK NTDSRCKARQ LELNERTCRC DKPRRHHHHH H | |
RUO | |
VEGFA | |
Unconjugated | |
Recombinant |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title