Learn More
R&D Systems™ Recombinant Human VSIG4 His-tag Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9678VS050
Catalog No. 96-78VS-050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human VSIG4 His-tag Protein is derived from NS0. The Recombinant Human VSIG4 His-tag Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Q9Y279-1
Lyophilized from a 0.2 μm filtered solution in PBS.
30 kDa
50 μg
Manual defrost freezer; Avoid repeated freeze-thaw cycles; 12 mo. from receipt, -20 to -70°C as supplied; 1 mo, 2 to 8°C under sterile conditions after reconstitution; 3 mo, -20 to -70°C under sterile conditions after reconstitution
<0.10 EU per 1 μg of the protein by the LAL method.
VSIG4
Reconstitute at 200 μg/mL in PBS.
Bioactivity
11326
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Mouse myeloma cell line, NS0-derived human VSIG4 protein Arg20-Pro283, with a C-terminal 6-His tag
RUO
Unconjugated
Human