Learn More
R&D Systems™ Recombinant Human VSIG4 Short Isoform His Tag Protein CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9816VS050
Catalog No. 98-16V-S050
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Human VSIG4 Short Isoform His Tag Protein is derived from CHO. The Recombinant Human VSIG4 Short Isoform His Tag Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Q9Y279-3
Lyophilized from a 0.2μm filtered solution in PBS.
20 kDa
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
CHO
<0.10 EU per 1μg of the protein by the LAL method.
Human
Bioactivity
11326
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily
50μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
Reconstitute at 200μg/mL in PBS.