Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant MERS-CoV Nucleocapsid His-tag Protein, CF
Sf21 Insect Cell Expressed, Over 95% Purity
Supplier: R&D Systems™ 10521CV100
This item is not returnable.
View return policy
Description
Bioassay data are not available.Specifications
YP_007188586.1 | |
Lyophilized from a 0.2 μm filtered solution in PBS. | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. | |
Spodoptera frugiperda, Sf 21 (baculovirus)-derived mers-cov Nucleocapsid protein Met1-Thr411, with a C-terminal 6-His tag | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Reconstitute at 100 μg/mL in PBS. |
Bioactivity | |
M.W.-observed: 49-55 kDa, under reducing conditions, M.W.-theoretical: 46 kDa | |
100 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Unconjugated | |
Nucleocapsid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title