Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse CAMK2N1 Protein

Highly purified. Generating reliable and reproducible results.

$280.50 - $783.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol VAMK2N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25957710 View Documents Novus Biologicals™
NBP25957710UG
10μg Each for $280.50
Add to cart
 
NBP25957750 View Documents Novus Biologicals™
NBP25957750UG
50μg Each for $783.00
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-78 of Mouse CAMK2N1 The Recombinant Mouse CAMK2N1 Protein is derived from E. coli. The Recombinant Mouse CAMK2N1 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

E.coli
RUO
VAMK2N1
55450
Recombinant Mouse CAMK2N1 Protein
Liquid. 20mM Tris-HCl (pH8.5) containing 1mM DTT, 0.1M NaCl, 10% glycerol
Unlabeled
0.5mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Calcium/Calmodulin Dependent Protein Kinase II Inhibitor 1, Calcium/Calmodulin-Dependent Protein Kinase II Inhibitor 1, CaMKII Inhibitory Protein Alpha, CaMKIINalpha, CaMKIIN-Alpha, PRO1489
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
10.9 kDa
Mouse

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit