Learn More
R&D Systems™ Recombinant Mouse CD6 His Tagged Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Manufacturer:  R&D Systems™ 9946CD100
Catalog No. 99-46CD-100
Add to cart
This item is not returnable. View return policy
Description
Description
The Recombinant Mouse CD6 His Tagged Protein is derived from NS0. The Recombinant Mouse CD6 His Tagged Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Q61003-1
Lyophilized from a 0.2 μm filtered solution in PBS.
42 kDa
100 μg
12 months from date of receipt, ≤ -20°C as supplied; 1 month, 2 to 8°C under sterile conditions after reconstitution; 3 months, ≤ -20°C under sterile conditions after reconstitution.
<0.10 EU per 1 μg of the protein by the LAL method.
CD6
Reconstitute at 500 μg/mL in PBS.
Bioactivity
923
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Mouse myeloma cell line, NS0-derived mouse CD6 protein Leu18-Gly396, with a C-Terminal 6-His tag
RUO
Unconjugated
Mouse