Learn More
Novus Biologicals™ Recombinant Mouse CNTF Animal-Free Protein
Highly purified and high bioactivity. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP2762111MG
 View more versions of this product
Catalog No. NBP2762111M
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
An un-tagged bioactive recombinant protein corresponding to Mouse CNTF
The Recombinant Mouse CNTF Animal-Free Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications
Specifications
1.4 mg/mL
1270
>95%, by SDS-PAGE
E. coli
CNTF
10 mM sodium phosphate, 150 mM sodium chloride, 10 mM DTT, 50% glycerol, pH 7.5
SDS-Page, Bioactivity
22.6 kDa
1 mg
Store at -20°C.
CNTF is a monomer, 22.6 kDa (198 aa)