Learn More
Novus Biologicals™ Recombinant Mouse Cyclophilin 40 Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
Manufacturer:  Novus Biologicals™ NBP2614515UG
 View more versions of this product
Catalog No. NBP2614515
Add to cart
Description
Description
A recombinant protein with a N-terminalHis-tag and corresponding to the amino acids 1-370 of Mouse Cyclophilin 40. The Recombinant Mouse Cyclophilin 40 Protein is derived from E. coli. The Recombinant Mouse Cyclophilin 40 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
1 mg/ml
20mM Tris-Hcl buffer (pH8.0) containing 10% glycerol, 1mM DTT
43.4 kDa
5 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
< 1.0 EU per 1μg of protein (determined by LAL method)
Cyclophilin 40
SDS-Page
5481
>90%, by SDS-PAGE
E. coli
RUO
PPID
Mouse