Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse EGF Isoform 1 Protein
SDP

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page, Bioactivity

$191.70 - $494.50

Specifications

Concentration 0.25 mg/ml
For Use With (Application) Bioactivity, SDS-PAGE
Formulation 20mM Tris-HCl(pH8.0) containing 0.1M NaCl, 2mM DTT, 10% glycerol
Gene ID (Entrez) 1950
Molecular Weight (g/mol) 8.6 kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP2619815
SDP
View Documents
Novus Biologicals™
NBP2619815UG
5 μg
Each for $191.70
Add to Cart
 
NBP26198120
SDP
View Documents
Novus Biologicals™
NBP26198120UG
20 μg
Each for $494.50
Add to Cart
 
Description

Description

A Biologically active recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 977-1029 of Mouse EGF The Recombinant Mouse EGF Isoform 1 Protein is derived from E. coli. The Recombinant Mouse EGF Isoform 1 Protein has been validated for the following applications: SDS-Page, Bioactivity.
Specifications

Specifications

0.25 mg/ml
20mM Tris-HCl(pH8.0) containing 0.1M NaCl, 2mM DTT, 10% glycerol
8.6 kDa
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
< 0.2 EU per 1μg of protein (determined by LAL method)
EGF
>95%, by SDS-PAGE
Bioactivity, SDS-PAGE
1950
E. coli
RUO
EGF
Mouse
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit