Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Mouse Enolase 2/Neuron-specific Enolase Protein

Highly purified. Generating reliable and reproducible results.

$129.70 - $351.00

Specifications

Source E.coli
Recombinant Yes
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Symbol ENO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP25953650 View Documents Novus Biologicals™
NBP25953650UG
50μg Each for $351.00
Add to cart
 
NBP25953610 View Documents Novus Biologicals™
NBP25953610UG
10μg Each for $129.70
Add to cart
 
Description

Description

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-434 of Mouse Enolase 2/Neuron-specific Enolase The Recombinant Mouse Enolase 2/Neuron-specific Enolase Protein is derived from E. coli. The Recombinant Mouse Enolase 2/Neuron-specific Enolase Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

E.coli
RUO
ENO2
2026
Recombinant Mouse Enolase 2/Neuron-specific Enolase Protein
Liquid. PBS (pH7.4) containing 10% glycerol
Unlabeled
1mg/mL
Yes
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
2-phospho-D-glycerate hydrolyase, 2-phospho-D-glycerate hydro-lyase, EC 4.2.1.11, Enolase 2, enolase 2 (gamma, neuronal), gamma-enolase, Neural enolase, neuron specific gamma enolase, neurone-specific enolase, Neuron-specific enolase, NSE
Recombinant Protein
>90%, by SDS-PAGE
SDS-Page
49.7 kDa

For Research Use Only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit